CAS RN 110-18-9
TEMED (Electrophoresis), Fisher BioReagents
CAS: 110-18-9 Molecular Formula: C6H16N2 Molecular Weight (g/mol): 116.208 InChI Key: KWYHDKDOAIKMQN-UHFFFAOYSA-N Synonym: temed,n,n,n',n'-tetramethylethylenediamine,tmeda,1,2-bis dimethylamino ethane,tetramethylethylenediamine,tetramethyldiaminoethane,tetrameen,propamine d,n1,n1,n2,n2-tetramethylethane-1,2-diamine,1,2-ethanediamine, n,n,n',n'-tetramethyl PubChem CID: 8037 ChEBI: CHEBI:32850 IUPAC Name: N,N,N',N'-tetramethylethane-1,2-diamine SMILES: CN(C)CCN(C)C
N,N,N',N'-Tetramethylethylenediamine, 99%, extra pure, Thermo Scientific™
N,N,N',N'-Tetramethylethylenediamine, 99.5%, purified by redistillation, AcroSeal™
CAS: 110-18-9 Molecular Formula: C6H16N2 Molecular Weight (g/mol): 116.21 InChI Key: KWYHDKDOAIKMQN-UHFFFAOYSA-N Synonym: temed,n,n,n',n'-tetramethylethylenediamine,tmeda,1,2-bis dimethylamino ethane,tetramethylethylenediamine,tetramethyldiaminoethane,tetrameen,propamine d,n1,n1,n2,n2-tetramethylethane-1,2-diamine,1,2-ethanediamine, n,n,n',n'-tetramethyl PubChem CID: 8037 ChEBI: CHEBI:32850 IUPAC Name: N,N,N',N'-tetramethylethane-1,2-diamine SMILES: CN(C)CCN(C)C
N,N,N',N'-Tetramethylethylenediamine, 99%
CAS: 110-18-9 Molecular Formula: C6H16N2 Molecular Weight (g/mol): 116.208 MDL Number: MFCD00008335 InChI Key: KWYHDKDOAIKMQN-UHFFFAOYSA-N Synonym: temed,n,n,n',n'-tetramethylethylenediamine,tmeda,1,2-bis dimethylamino ethane,tetramethylethylenediamine,tetramethyldiaminoethane,tetrameen,propamine d,n1,n1,n2,n2-tetramethylethane-1,2-diamine,1,2-ethanediamine, n,n,n',n'-tetramethyl PubChem CID: 8037 ChEBI: CHEBI:32850 IUPAC Name: N,N,N',N'-tetramethylethane-1,2-diamine SMILES: CN(C)CCN(C)C
N,N,N',N'-Tetramethylethylenediamine, 99%, extra pure
CAS: 110-18-9 Molecular Formula: C6H16N2 Molecular Weight (g/mol): 116.21 InChI Key: KWYHDKDOAIKMQN-UHFFFAOYSA-N Synonym: temed,n,n,n',n'-tetramethylethylenediamine,tmeda,1,2-bis dimethylamino ethane,tetramethylethylenediamine,tetramethyldiaminoethane,tetrameen,propamine d,n1,n1,n2,n2-tetramethylethane-1,2-diamine,1,2-ethanediamine, n,n,n',n'-tetramethyl PubChem CID: 8037 ChEBI: CHEBI:32850 IUPAC Name: N,N,N',N'-tetramethylethane-1,2-diamine SMILES: CN(C)CCN(C)C
Thermo Scientific Chemicals N,N,N',N'-Tetramethylethylenediamine, 99%, for biochemistry, For electrophoresis
CAS: 110-18-9 Molecular Formula: C6H16N2 Molecular Weight (g/mol): 116.21 MDL Number: MFCD00008335 InChI Key: KWYHDKDOAIKMQN-UHFFFAOYSA-N Synonym: temed,n,n,n',n'-tetramethylethylenediamine,tmeda,1,2-bis dimethylamino ethane,tetramethylethylenediamine,tetramethyldiaminoethane,tetrameen,propamine d,n1,n1,n2,n2-tetramethylethane-1,2-diamine,1,2-ethanediamine, n,n,n',n'-tetramethyl PubChem CID: 8037 ChEBI: CHEBI:32850 IUPAC Name: N,N,N',N'-tetramethylethane-1,2-diamine SMILES: CN(C)CCN(C)C
Thermo Scientific Chemicals N,N,N',N'-Tetramethylethylenediamine, Electrophoresis Grade
CAS: 110-18-9 Molecular Formula: C6H16N2 Molecular Weight (g/mol): 116.208 MDL Number: MFCD00008335 InChI Key: KWYHDKDOAIKMQN-UHFFFAOYSA-N Synonym: temed,n,n,n',n'-tetramethylethylenediamine,tmeda,1,2-bis dimethylamino ethane,tetramethylethylenediamine,tetramethyldiaminoethane,tetrameen,propamine d,n1,n1,n2,n2-tetramethylethane-1,2-diamine,1,2-ethanediamine, n,n,n',n'-tetramethyl PubChem CID: 8037 ChEBI: CHEBI:32850 IUPAC Name: N,N,N',N'-tetramethylethane-1,2-diamine SMILES: CN(C)CCN(C)C
N,N,N',N'-Tetramethyl-Ethylenediamine, >99%, MP Biomedicals™
CAS: 110-18-9 Molecular Formula: C6H16N2 Molecular Weight (g/mol): 116.208 InChI Key: KWYHDKDOAIKMQN-UHFFFAOYSA-N Synonym: temed,n,n,n',n'-tetramethylethylenediamine,tmeda,1,2-bis dimethylamino ethane,tetramethylethylenediamine,tetramethyldiaminoethane,tetrameen,propamine d,n1,n1,n2,n2-tetramethylethane-1,2-diamine,1,2-ethanediamine, n,n,n',n'-tetramethyl PubChem CID: 8037 ChEBI: CHEBI:32850 IUPAC Name: N,N,N',N'-tetramethylethane-1,2-diamine SMILES: CN(C)CCN(C)C
N,N,N',N'-Tetramethylethylenediamine, Electrophoresis Grade, MP Biomedicals
CAS: 110-18-9 Molecular Formula: C6H16N2 Molecular Weight (g/mol): 116.208 InChI Key: KWYHDKDOAIKMQN-UHFFFAOYSA-N Synonym: temed,n,n,n',n'-tetramethylethylenediamine,tmeda,1,2-bis dimethylamino ethane,tetramethylethylenediamine,tetramethyldiaminoethane,tetrameen,propamine d,n1,n1,n2,n2-tetramethylethane-1,2-diamine,1,2-ethanediamine, n,n,n',n'-tetramethyl PubChem CID: 8037 ChEBI: CHEBI:32850 IUPAC Name: N,N,N',N'-tetramethylethane-1,2-diamine SMILES: CN(C)CCN(C)C
TEMED, Approx. 99%, Molecular Biology Reagent, MP Biomedicals™
CAS: 110-18-9 Molecular Formula: C6H16N2 Molecular Weight (g/mol): 116.208 MDL Number: MFCD00008335 InChI Key: KWYHDKDOAIKMQN-UHFFFAOYSA-N Synonym: temed,n,n,n',n'-tetramethylethylenediamine,tmeda,1,2-bis dimethylamino ethane,tetramethylethylenediamine,tetramethyldiaminoethane,tetrameen,propamine d,n1,n1,n2,n2-tetramethylethane-1,2-diamine,1,2-ethanediamine, n,n,n',n'-tetramethyl PubChem CID: 8037 ChEBI: CHEBI:32850 IUPAC Name: N,N,N',N'-tetramethylethane-1,2-diamine SMILES: CN(C)CCN(C)C
N,N,Nft.,Nft.-Tetramethylethylenediamine, Molecular biology reagent grade, MP Biomedicals™
CAS: 110-18-9 Molecular Formula: C6H16N2 Molecular Weight (g/mol): 116.208 MDL Number: MFCD00008335 InChI Key: KWYHDKDOAIKMQN-UHFFFAOYSA-N Synonym: temed,n,n,n',n'-tetramethylethylenediamine,tmeda,1,2-bis dimethylamino ethane,tetramethylethylenediamine,tetramethyldiaminoethane,tetrameen,propamine d,n1,n1,n2,n2-tetramethylethane-1,2-diamine,1,2-ethanediamine, n,n,n',n'-tetramethyl PubChem CID: 8037 ChEBI: CHEBI:32850 IUPAC Name: N,N,N',N'-tetramethylethane-1,2-diamine SMILES: CN(C)CCN(C)C
N,N,N',N'-Tetramethylethylenediamine 98.0+%, TCI America™
CAS: 110-18-9 Molecular Formula: C6H16N2 Molecular Weight (g/mol): 116.208 MDL Number: MFCD00008335 InChI Key: KWYHDKDOAIKMQN-UHFFFAOYSA-N Synonym: temed,n,n,n',n'-tetramethylethylenediamine,tmeda,1,2-bis dimethylamino ethane,tetramethylethylenediamine,tetramethyldiaminoethane,tetrameen,propamine d,n1,n1,n2,n2-tetramethylethane-1,2-diamine,1,2-ethanediamine, n,n,n',n'-tetramethyl PubChem CID: 8037 ChEBI: CHEBI:32850 IUPAC Name: N,N,N',N'-tetramethylethane-1,2-diamine SMILES: CN(C)CCN(C)C
N,N,N',N'-Tetramethylethylenediamine 98.0+%, TCI America™
CAS: 110-18-9 Molecular Formula: C6H16N2 Molecular Weight (g/mol): 116.208 MDL Number: MFCD00008335 InChI Key: KWYHDKDOAIKMQN-UHFFFAOYSA-N Synonym: temed,n,n,n',n'-tetramethylethylenediamine,tmeda,1,2-bis dimethylamino ethane,tetramethylethylenediamine,tetramethyldiaminoethane,tetrameen,propamine d,n1,n1,n2,n2-tetramethylethane-1,2-diamine,1,2-ethanediamine, n,n,n',n'-tetramethyl PubChem CID: 8037 ChEBI: CHEBI:32850 IUPAC Name: N,N,N',N'-tetramethylethane-1,2-diamine SMILES: CN(C)CCN(C)C
N,N,N',N'-Tetramethylethylenediamine (TEMED), Hoefer™
CAS: 110-18-9 Molecular Formula: C6H16N2 Molecular Weight (g/mol): 116.208 InChI Key: KWYHDKDOAIKMQN-UHFFFAOYSA-N Synonym: temed,n,n,n',n'-tetramethylethylenediamine,tmeda,1,2-bis dimethylamino ethane,tetramethylethylenediamine,tetramethyldiaminoethane,tetrameen,propamine d,n1,n1,n2,n2-tetramethylethane-1,2-diamine,1,2-ethanediamine, n,n,n',n'-tetramethyl PubChem CID: 8037 ChEBI: CHEBI:32850 IUPAC Name: N,N,N',N'-tetramethylethane-1,2-diamine SMILES: CN(C)CCN(C)C
- 1
- 2