missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,N',N'-Tetramethylethylenediamine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | -55°C |
| Color | Yellow |
| Boiling Point | 122°C |
| Quantity | 5 g |
| UN Number | 2372 |
| Formula Weight | 116.21 |
| Percent Purity | ≥98.0% (GC,T) |
| Grade | Electrophoresis |
| Physical Form | Liquid |
| Chemical Name or Material | N,N,N′,N′-Tetramethylethylenediamine [for Electrophoresis] |
Safety and Handling
| EINECSNumber | (2)-0155 |
| RTECSNumber | KV7175000 |
| TSCA | Yes |