missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-Ethylenediamine, >99%, MP Biomedicals™

Catalog No. ICN805614
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN805614 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN805614 Supplier MP Biomedicals Inc Supplier No. 04805614

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used as a catalyst for polyacrylamide gel formation to promote acrylamide polymerization.

Chemical Identifiers

CAS 110-18-9
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.208
InChI Key KWYHDKDOAIKMQN-UHFFFAOYSA-N
Synonym temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
PubChem CID 8037
ChEBI CHEBI:32850
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine
SMILES CN(C)CCN(C)C

Specifications

Assay Percent Range >99%
Quantity 25 mL
Formula Weight 116.2
Percent Purity ≥99%
Grade Electrophoresis
Physical Form Liquid
Chemical Name or Material N,N,N',N'-Tetramethyl-Ethylenediamine
Recommended Storage Store at room temperature