missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ N,N,N',N'-Tetramethylethylenediamine, 99%, for biochemistry, For electrophoresis

Catalog No. AC420580025
Change view
Click to view available options
2.5kg; Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC420580025 2.5kg; Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC420580025 Supplier Thermo Scientific™ Supplier No. 420580025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 110-18-9
CAS Min % 98.5
CAS Max % 100.0
Density 0.7700g/mL
Boiling Point 120.0°C to 122.0°C
Flash Point 17°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic Bottle
Molecular Formula C6H16N2
Refractive Index 1.4169 to 1.4189
Linear Formula (CH3)2NCH2CH2N(CH3)2
Quantity 2.5kg
MDL Number MFCD00008335
Beilstein 04, 250
Fieser 02,403; 03,284; 04,485; 05,652; 06,576; 07,358; 11,340; 12,477; 14,68; 15,63; 16,55
Specific Gravity 0.77
Merck Index 15, 9277
Synonym temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
Solubility Information (10% in water) Clear and no haze
InChI Key KWYHDKDOAIKMQN-UHFFFAOYSA-N
SMILES CN(C)CCN(C)C
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine
Molecular Weight (g/mol) 116.21
PubChem CID 8037
ChEBI CHEBI:32850
Formula Weight 116.21
Percent Purity 99%
Grade Biochemistry
Chemical Name or Material N,N,N',N'-Tetramethylethylenediamine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-744-6
RTECSNumber KV7175000
TSCA TSCA

RUO – Research Use Only