missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N,N,N',N'-Tetramethylethylenediamine, Electrophoresis Grade

Catalog No. AAJ63734AE
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63734AE 100 mL
AAJ63734AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63734AE Supplier Thermo Scientific Chemicals Supplier No. J63734AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 110-18-9 | C6H16N2 | 116.208 g/mol

N,N,N',N'-Tetramethylethylenediamine is used as polymerization accelerator in gel electrophoresis, solvent and oxidizing reagent. N,N,N',N'-Tetramethylethylenediamine is used as anti-hyperlipidemic, plant growth stimulator, pathogen growth inhibitor, confers plant disease resistance. TEMED is used with ammonium persulfate to catalyze the polymerization of acrylamide when making polyacrylamide gels, used in gel electrophoresis. TMEDA is widely employed as a ligand for metal ions. TEMED can also be a component of Hypergolic propellants. Tetramethylethylenediamine (TEMED) is an essential catalyst for polyacrylamide gel polymerization. TEMED is frequently used with another catalyst, APS, for the preparation of polyacrylamide gels for protein and nucleic acid analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N,N′,N′-Tetramethylethylenediamine is used as polymerization accelerator in gel electrophoresis, solvent and oxidizing reagent. N,N,N′,N′-Tetramethylethylenediamine is used as anti-hyperlipidemic, plant growth stimulator, pathogen growth inhibitor, confers plant disease resistance. TEMED is used with ammonium persulfate to catalyze the polymerization of acrylamide when making polyacrylamide gels, used in gel electrophoresis. TMEDA is widely employed as a ligand for metal ions. TEMED can also be a component of Hypergolic propellants. Tetramethylethylenediamine (TEMED) is an essential catalyst for polyacrylamide gel polymerization. TEMED is frequently used with another catalyst, APS, for the preparation of polyacrylamide gels for protein and nucleic acid analysis.

Solubility
Soluble in water 10 mg/mL at 20°C.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides, copper, mercury, water, moisture.

Chemical Identifiers

CAS 110-18-9
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.208
MDL Number MFCD00008335
InChI Key KWYHDKDOAIKMQN-UHFFFAOYSA-N
Synonym temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
PubChem CID 8037
ChEBI CHEBI:32850
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine
SMILES CN(C)CCN(C)C

Specifications

Melting Point -55°C
Color Colorless
Density 0.775
Boiling Point 120°C to 122°C
Flash Point 18°C (154°F)
Odor Unpleasant
Refractive Index 1.418
Linear Formula (CH3)2NCH2CH2N(CH3)2
Quantity 100 mL
UN Number UN2372
Beilstein 1732991
Sensitivity Hygroscopic
Merck Index 14,9134
Solubility Information Soluble in water 10mg/mL at 20°C.
Formula Weight 116.21
Grade Electrophoresis
Physical Form Liquid
Chemical Name or Material N,N,N',N'-Tetramethylethylenediamine
Show More Show Less
Hazard Category H225-H302+H332-H314-H335
Hazard Statement GHS H Statement
H225-H314-H302-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1,2-DI-(DIMETHYLAMINO)ETHANE
EINECSNumber 203-744-6
RTECSNumber KV7175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only