missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethylethylenediamine, 99%

Catalog No. AAA12536AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA12536AP 500 mL
AAA12536AE 100 mL
AAA125360F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA12536AP Supplier Thermo Scientific Chemicals Supplier No. A12536AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 110-18-9 | C6H16N2 | 116.208 g/mol

N,N,N',N'-Tetramethylethylenediamine is used with ammonium persulfate to catalyze the polymerization of acrylamide and bis-acrylamide to get polyacrylamide gels. It is also used for the separation of proteins or nucleic acids. Further, it is employed as a ligand for metal ions like zinc and copper. It is actively involved in the formation of anionic organometallic complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N,N′,N′-Tetramethylethylenediamine is used with ammonium persulfate to catalyze the polymerization of acrylamide and bis-acrylamide to get polyacrylamide gels. It is also used for the separation of proteins or nucleic acids. Further, it is employed as a ligand for metal ions like zinc and copper. It is actively involved in the formation of anionic organometallic complex.

Solubility
Miscible with water.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, carbon dioxide and copper.

Chemical Identifiers

CAS 110-18-9
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.208
MDL Number MFCD00008335
InChI Key KWYHDKDOAIKMQN-UHFFFAOYSA-N
Synonym temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
PubChem CID 8037
ChEBI CHEBI:32850
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine
SMILES CN(C)CCN(C)C

Specifications

Melting Point -55°C
Density 0.775
Boiling Point 120°C to 122°C
Flash Point 18°C (62°F)
Odor Unpleasant
Assay Percent Range 99%
Refractive Index 1.418
Linear Formula (CH3)2NCH2CH2N(CH3)2
Quantity 500 mL
UN Number UN2372
Beilstein 1732991
Sensitivity Air sensitive; Hygroscopic
Solubility Information Miscible with water.
Formula Weight 116.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N,N,N',N'-Tetramethylethylenediamine
Show More Show Less
Hazard Category H225-H302+H332-H314-H335
Hazard Statement GHS H Statement
H225-H314-H302-H332
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1,2-DI-(DIMETHYLAMINO)ETHANE
EINECSNumber 203-744-6
RTECSNumber KV7175000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only