missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich N,N,N',N'-Tetramethylethylenediamine

Catalog No. 411019500ML
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
411019500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 411019500ML Supplier Sigma Aldrich Supplier No. 411019500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N,N,N',N'-Tetramethylethylenediamine

Specifications

CAS 110-18-9
Melting Point -55°C (lit.)
Density 0.775 g/mL (at 20°C (literature))
Boiling Point 120°C to 122°C (lit.)
Molecular Formula C6H16N2
Refractive Index n20/D 1.4179 (literature)
Linear Formula (CH3)2 NCH2CH2N(CH3)2
MDL Number MFCD00008335
Quantity 500 mL
Synonym 1,2-Bis(dimethylamino)ethane; TEMED; TMEDA
Molecular Weight (g/mol) 116.2
Percent Purity ≥99.5%
Show More Show Less