CAS RN 108-88-3
Toluene (Certified ACS), Fisher Chemical™
CAS: 108-88-3 Molecular Formula: C7H8 Molecular Weight (g/mol): 92.14 MDL Number: MFCD00008512 InChI Key: YXFVVABEGXRONW-UHFFFAOYSA-N Synonym: methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene PubChem CID: 1140 ChEBI: CHEBI:17578 IUPAC Name: toluene SMILES: CC1=CC=CC=C1
Toluene (HPLC), Fisher Chemical™
CAS: 108-88-3 Molecular Formula: C7H8 Molecular Weight (g/mol): 92.14 MDL Number: MFCD00008512 InChI Key: YXFVVABEGXRONW-UHFFFAOYSA-N Synonym: methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene PubChem CID: 1140 ChEBI: CHEBI:17578 IUPAC Name: toluene SMILES: CC1=CC=CC=C1
Toluene, Optima™, Fisher Chemical™
CAS: 108-88-3 Molecular Formula: C7H8 Molecular Weight (g/mol): 92.14 MDL Number: MFCD00008512 InChI Key: YXFVVABEGXRONW-UHFFFAOYSA-N Synonym: methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene PubChem CID: 1140 ChEBI: CHEBI:17578 IUPAC Name: toluene SMILES: CC1=CC=CC=C1
Toluene, PRF, Meets requirements of ASTM D2699 and D2700, Solstice
CAS: 108-88-3 Molecular Formula: C7H8 Molecular Weight (g/mol): 92.14 MDL Number: MFCD00008512 InChI Key: YXFVVABEGXRONW-UHFFFAOYSA-N Synonym: methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene PubChem CID: 1140 ChEBI: CHEBI:17578 IUPAC Name: toluene SMILES: CC1=CC=CC=C1
Toluene, Optima™ for HPLC and GC, Fisher Chemical
Toluene, 99%
CAS: 108-88-3 Molecular Formula: C7H8 Molecular Weight (g/mol): 92.14 MDL Number: MFCD00008512 InChI Key: YXFVVABEGXRONW-UHFFFAOYSA-N Synonym: methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene PubChem CID: 1140 ChEBI: CHEBI:17578 IUPAC Name: toluene SMILES: CC1=CC=CC=C1
Toluene, ACS, 99.5% min
CAS: 108-88-3 Molecular Formula: C7H8 Molecular Weight (g/mol): 92.14 MDL Number: MFCD00008512 InChI Key: YXFVVABEGXRONW-UHFFFAOYSA-N Synonym: methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene PubChem CID: 1140 ChEBI: CHEBI:17578 IUPAC Name: toluene SMILES: CC1=CC=CC=C1
Toluene, 99.85%, for HPLC
CAS: 108-88-3 Molecular Formula: C7H8 Molecular Weight (g/mol): 92.14 MDL Number: MFCD00008512 InChI Key: YXFVVABEGXRONW-UHFFFAOYSA-N Synonym: methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene PubChem CID: 1140 ChEBI: CHEBI:17578 IUPAC Name: toluene SMILES: CC1=CC=CC=C1
Toluene, Semiconductor Grade, 99% min
CAS: 108-88-3 Molecular Formula: C7H8 Molecular Weight (g/mol): 92.14 MDL Number: MFCD00008512 InChI Key: YXFVVABEGXRONW-UHFFFAOYSA-N Synonym: methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene PubChem CID: 1140 ChEBI: CHEBI:17578 IUPAC Name: toluene SMILES: CC1=CC=CC=C1
Toluene, 99.5%, for spectroscopy ACS
CAS: 108-88-3 Molecular Formula: C7H8 Molecular Weight (g/mol): 92.14 MDL Number: MFCD00008512 InChI Key: YXFVVABEGXRONW-UHFFFAOYSA-N Synonym: methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene PubChem CID: 1140 ChEBI: CHEBI:17578 IUPAC Name: toluene SMILES: CC1=CC=CC=C1
Toluene, Laboratory Reagent, ≥99.3%, Solstice
CAS: 108-88-3 Molecular Formula: C7H8 Molecular Weight (g/mol): 92.14 MDL Number: MFCD00008512 InChI Key: YXFVVABEGXRONW-UHFFFAOYSA-N Synonym: methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene PubChem CID: 1140 ChEBI: CHEBI:17578 IUPAC Name: toluene SMILES: CC1=CC=CC=C1
Toluene, ACS Reagent, for HPLC, Spectrophotometry, meets analytical spec. of USP, >99.8%, Solstice
CAS: 108-88-3 Molecular Formula: C7H8 Molecular Weight (g/mol): 92.14 MDL Number: MFCD00008512 InChI Key: YXFVVABEGXRONW-UHFFFAOYSA-N Synonym: methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene PubChem CID: 1140 ChEBI: CHEBI:17578 IUPAC Name: toluene SMILES: CC1=CC=CC=C1
Toluene, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.8%, Solstice
CAS: 108-88-3 Molecular Formula: C7H8 Molecular Weight (g/mol): 92.14 MDL Number: MFCD00008512 InChI Key: YXFVVABEGXRONW-UHFFFAOYSA-N Synonym: methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene PubChem CID: 1140 ChEBI: CHEBI:17578 IUPAC Name: toluene SMILES: CC1=CC=CC=C1
Toluene, HPLC Grade, 99.7% min
CAS: 108-88-3 Molecular Formula: C7H8 Molecular Weight (g/mol): 92.14 MDL Number: MFCD00008512 InChI Key: YXFVVABEGXRONW-UHFFFAOYSA-N Synonym: methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene PubChem CID: 1140 ChEBI: CHEBI:17578 IUPAC Name: toluene SMILES: CC1=CC=CC=C1
Toluene, 99.5%, ACS reagent, meets the requirements of Reag. Ph.Eur.
CAS: 108-88-3 Molecular Formula: C7H8 Molecular Weight (g/mol): 92.14 MDL Number: MFCD00008512 InChI Key: YXFVVABEGXRONW-UHFFFAOYSA-N Synonym: methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene PubChem CID: 1140 ChEBI: CHEBI:17578 IUPAC Name: toluene SMILES: CC1=CC=CC=C1