missing translation for 'onlineSavingsMsg'
Learn More

Toluene, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111017857
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017857 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017857 Supplier Merck Emd Millipore Supplier No. PHR13111G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Toluene is a non-polar organic solvent and methyl derivative of benzene. As per USP and EP, it belongs to the Class 2 category of organic solvents and has limited application in the synthesis of pharmaceuticals, due to its moderate toxicity.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 108-88-3
Vapor Pressure 26 mmHg (25°C)
Vapor Density 3.2 (vs. air)
Density 0.865 g/mL (at 25°C)
Physical Form Neat
Linear Formula C6H5CH3
MDL Number MFCD00008512
Melting Point -93°C
Boiling Point 110°C to 111°C
Quantity 1 g
Formula Weight 92.14
Grade Certified Reference Material
Show More Show Less