missing translation for 'onlineSavingsMsg'
Learn More

Toluene, 99.85%, for pesticide residue analysis

Catalog No. AC332070010
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC332070010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC332070010 Supplier Thermo Fisher Scientific Formerly Acros Organics Supplier No. 332070010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-88-3 | C7H8 | 92.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For residue analysis

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -95.0°C
Color Colorless to Yellow
Acidity ≤0.003% (as HCl)
Density 0.8660g/mL
Quantity 1 L
Flash Point 4°C
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4968
Residue after Evaporation 0.0005 % max.
Specific Gravity 0.866
Assay Percent Range 99.85%
Vapor Pressure Vapor Pressure: 29mbar at 20°C
Linear Formula C6H5CH3
Viscosity 0.6 mPa.s (20°C)
Water 0.02% Max. (K.F.)
Beilstein 05, 280
Merck Index 15, 9688
Solubility Information Solubility in water: almost insoluble (0.5g/L at 20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, acetone,, ether, petroleum ether, benzene, ch2cl2, dmf, gla, cial acetic acid, chcl3, ccl4.
Boiling Point 111.0°C
Formula Weight 92.14
Percent Purity 99.85%
Grade Pesticide
Chemical Name or Material Toluene, For Pesticide Residue Analysis
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. May be fatal if swallowed and enters airways. Causes skin irritation. Suspected of damaging the unborn child. Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 203-625-9
RTECSNumber XS5250000
TSCA TSCA

RUO – Research Use Only