missing translation for 'onlineSavingsMsg'
Learn More

Toluene (HPLC), Fisher Chemical™

Catalog No. T2904
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Safe-Cote™ Amber Glass 
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
T2904 4 L Glass Bottle
T290SK4 4 L Safe-Cote™ Amber Glass 
T2901 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T2904 Supplier Thermo Fisher Scientific Formerly Fairlawn Supplier No. BPT2904
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-88-3 | C7H8 | 92.14 g/mol

  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested by GC-ECD to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Also meets ACS specifications
  • Intended for research use only

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -95°C
Absorbance 1 AU max. at 286nm
Color Colorless
Density 0.866g/cm³
Quantity 4 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4930 to 1.4950 (at 25°C)
Residue after Evaporation 10ppm max.
Assay Percent Range 99.8% min by GC
Vapor Pressure 29mbar at 20°C
Viscosity 0.6 mPa/s at 20°C
Water 0.02% max.
Boiling Point 111°C
Formula Weight 92.14
Percent Purity ≥99.8%
Grade Mobile phase for HPLC applications
Chemical Name or Material Toluene, Also meets ACS specifications
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Category Signal Word: DANGER!
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0
DOTInformation DOT Class 3, : Flammable Liquid