accessibility menu, dialog, popup

UserName

CH3H3COCH3O

CAS RN 108-21-4

IUPAC Name: propan-2-yl acetate
Synonyms: isopropyl acetate
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: JMMWKPVZQRWMSS-UHFFFAOYSA-N
SMILES: CC(C)OC(C)=O
Percent Purity 
  • (3)
  • (2)
  • (2)
  • (4)
  • (3)
  • (5)
Boiling Point 
  • (1)
  • (3)
  • (2)
  • (8)
  • (4)
Quantity 
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (8)
  • (6)
  • (2)
Grade 
  • (2)
  • (2)
  • (1)
  • (6)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (3)
  • (4)
  • (1)
  • (2)
  • (11)
  • (3)
  • (2)
  • (2)
  • (4)
  • (3)
  • (5)
  • (1)
  • (3)
  • (2)
  • (8)
  • (4)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (8)
  • (6)
  • (2)
  • (2)
  • (2)
  • (1)
  • (6)
  • (4)
  • (10)
  • (4)
  • (1)
  • (1)