missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Acetate, Spex CertiPrep

Catalog No. S2305
Change view
Click to view available options
Quantity:
1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S2305 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S2305 Supplier SPEX CertiPrep Group LLC Supplier No. S2305

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Isopropyl Acetate Organic Single in Methanol-P&T, 1 mL, 1000 μg/mL, SPEX CertiPrep™

  • Isopopyl Acetate
  • CAS #108-21-4
  • 1 mL volume

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Chemical Identifiers

CAS 108-21-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
InChI Key JMMWKPVZQRWMSS-UHFFFAOYSA-N
PubChem CID 7915
IUPAC Name propan-2-yl acetate
SMILES CC(C)OC(=O)C

Specifications

Packaging Ampule
Physical Form Liquid
Technique AA, ICP
Type Certified Reference Material
Quantity 1.2 mL
Chemical Name or Material Isopropyl Acetate