missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl acetate, Analytical Standard, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019332
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019332 5 mL
111018755 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019332 Supplier Merck Emd Millipore Supplier No. 908715MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Isopropyl acetate is an isopropyl ester of acetic acid. It participates in the mesoporous Al-MCM-41 (Si/Al = 55 and 104) and Al, Zn-MCM-41 (Si/(Al+Zn) = 52) molecular sieves catalyzed alkylation of m-cresol. It is widely used for the incorporation of aroma to various cosmetics and food products. Vapor-liquid equilibria for its binary mixture with CO2 at higher pressures has been evaluated. It is a colorless, flammable liquid, having a pleasant fruity type of odor.

Specifications

CAS 108-21-4
Vapor Pressure 47 mmHg (20°C)
Vapor Density 3.5 (vs. air)
Density 0.872 g/mL (at 25°C)
Physical Form Neat
Linear Formula CH3COOCH(CH3)2
MDL Number MFCD00008877
Melting Point -73°C
Boiling Point 85°C to 91°C
Synonym Acetic acid isopropyl ester
Quantity 5 mL
Formula Weight 102.13
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less