missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl acetate, 99+%

Catalog No. AAL09337AU
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL09337AU 1000 mL
AAL09337AK 250 mL
AAL093370F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL09337AU Supplier Thermo Scientific Chemicals Supplier No. L09337AU

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-21-4 | C5H10O2 | 102.133 g/mol

Isopropyl acetate is a solvent in chemical industry, especially for cellulose, plastics, waxes, resins, gums, paints, oil and fats. and also as flavoring agent. It is an active component of perfumes and printing inks. It is also employed as an extractant for the preparation of antibiotics, vitamins and hormones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropyl acetate is a solvent in chemical industry, especially for cellulose, plastics, waxes, resins, gums, paints, oil and fats. and also as flavoring agent. It is an active component of perfumes and printing inks. It is also employed as an extractant for the preparation of antibiotics, vitamins and hormones.

Solubility
Slightly miscible with cold water and methanol.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and bases.

Chemical Identifiers

CAS 108-21-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00008877
InChI Key JMMWKPVZQRWMSS-UHFFFAOYSA-N
Synonym isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl ester
PubChem CID 7915
IUPAC Name propan-2-yl acetate
SMILES CC(C)OC(=O)C

Specifications

Melting Point -73°C
Density 0.87
Boiling Point 87°C to 89°C
Flash Point 4°C (39°F)
Odor Ester-like
Refractive Index 1.377
Linear Formula CH3CO2CH(CH3)2
Quantity 1000 mL
UN Number UN1220
Beilstein 1740761
Sensitivity Moisture sensitive
Merck Index 14,5205
Solubility Information Slightly miscible with cold water and methanol.
Formula Weight 102.13
Percent Purity ≥99%
Chemical Name or Material Isopropyl acetate
Show More Show Less
Hazard Category H225-H319-H336
Hazard Statement GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPYL ACETATE
EINECSNumber 203-561-1
RTECSNumber AI4930000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only