missing translation for 'onlineSavingsMsg'
Learn More
Learn More
iso-Propyl Acetate, 98%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Boiling Point | 89°C/192.2°F |
| Packaging | Amber Glass Bottle |
| Quantity | 500 mL |
| Percent Purity | 0.98 |
| Grade | Ungraded |
| Physical Form | Viscous Liquid or Low Melting Solid |
| Chemical Name or Material | iso-Propyl Acetate |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.