missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl acetate, 99+%, pure

Catalog No. AC180800250
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC180800250 25 L
AC180800010 1 L
AC180800025 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC180800250 Supplier Thermo Scientific Chemicals Supplier No. 180800250

missing translation for 'validMainframeMsg'

CAS: 108-21-4 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-21-4
MDL Number MFCD00008877
InChI Key JMMWKPVZQRWMSS-UHFFFAOYSA-N
Synonym isopropyl acetate, 2-propyl acetate, 2-acetoxypropane, acetic acid, 1-methylethyl ester, isopropyl ethanoate, isopropylacetat, paracetat, isopropylacetaat, 1-methylethyl acetate, acetic acid, isopropyl ester
PubChem CID 7915
IUPAC Name propan-2-yl acetate
SMILES CC(C)OC(=O)C

Specifications

Melting Point -73.0°C
Color Colorless
Density 0.8720g/mL
Boiling Point 88.8°C
Flash Point 4°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Metal drum
Molecular Formula C5H10O2
Refractive Index 1.3760 to 1.3780
Linear Formula CH3CO2CH(CH3)2
Quantity 25 L
Residue after Evaporation 0.01% max.
Beilstein 02, 130
Specific Gravity 0.872
Merck Index 15, 5251
Solubility Information Solubility in water: 31g/L (20°C). Other solubilities: soluble in most common organic solvents
Molecular Weight (g/mol) 102.13
Viscosity 0.49 mPa.s (25°C)
Formula Weight 102.13
Percent Purity 99+%
Grade Pure
Physical Form Liquid
Chemical Name or Material Isopropyl acetate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-561-1

RUO – Research Use Only