accessibility menu, dialog, popup

UserName

CAS RN 1073-70-7

IUPAC Name: hydrogen (4-chlorophenyl)hydrazine chloride
Synonyms: hydrogen 4-chlorophenylhydrazine chloride
Molecular Weight (g/mol): 179.04
Molecular Formula: C6H8Cl2N2
InChi Key: YQVZREHUWCCHHX-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (3)
  • (1)
  • (2)
  • (4)