missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylhydrazine hydrochloride, 97%

Catalog No. AC404950250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC404950250 25 g
AC404950050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC404950250 Supplier Thermo Scientific Chemicals Supplier No. 404950250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1073-70-7 | C6H7ClN2·HCl | 179.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1073-70-7
Molecular Formula C6H7ClN2·HCl
Molecular Weight (g/mol) 179.05
MDL Number MFCD00012943
InChI Key YQVZREHUWCCHHX-UHFFFAOYSA-N
Synonym 4-chlorophenylhydrazine hydrochloride, 4-chlorophenyl hydrazine hydrochloride, p-chlorophenylhydrazine hydrochloride, p-chlorophenylhydrazine hcl, 4-chlorophenylhydrazine hcl, 1-4-chlorophenyl hydrazine hydrochloride, 4-chloro-phenyl-hydrazine hydrochloride, chlorophenylhydrazine 4-hcl, hydrazine, 4-chlorophenyl-, monohydrochloride, 4-chlorophenylhydrazine, chloride
PubChem CID 71600
IUPAC Name (4-chlorophenyl)hydrazine;hydrochloride
SMILES C1=CC(=CC=C1NN)Cl.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 203.0°C
Color Pink to White
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 96.0 to 104% (Argentometry)
Packaging Glass bottle
Linear Formula NHNH2C6H4Cl·HCl
Quantity 25 g
Beilstein 15,I,106
Formula Weight 179.05
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorophenylhydrazine hydrochloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove
EINECSNumber 214-030-9

RUO – Research Use Only