missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylhydrazine Hydrochloride 98.0+%, TCI America™

Catalog No. C025725G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C025725G 25 g
C0257250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C025725G Supplier TCI America Supplier No. C025725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1073-70-7
Molecular Formula C6H8Cl2N2
Molecular Weight (g/mol) 179.044
MDL Number MFCD00012943
InChI Key YQVZREHUWCCHHX-UHFFFAOYSA-N
Synonym 4-chlorophenylhydrazine hydrochloride, 4-chlorophenyl hydrazine hydrochloride, p-chlorophenylhydrazine hydrochloride, p-chlorophenylhydrazine hcl, 4-chlorophenylhydrazine hcl, 1-4-chlorophenyl hydrazine hydrochloride, 4-chloro-phenyl-hydrazine hydrochloride, chlorophenylhydrazine 4-hcl, hydrazine, 4-chlorophenyl-, monohydrochloride, 4-chlorophenylhydrazine, chloride
PubChem CID 71600
IUPAC Name (4-chlorophenyl)hydrazine;hydrochloride
SMILES C1=CC(=CC=C1NN)Cl.Cl

Specifications

Quantity 25 g
Formula Weight 179.04
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorophenylhydrazine Hydrochloride