missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylhydrazine hydrochloride, 97%

Catalog No. AAA1451822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1451822 100 g
AAA1451806 5 g
AAA1451814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1451822 Supplier Thermo Scientific Chemicals Supplier No. A1451822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1073-70-7 | C6H8Cl2N2 | 179.044 g/mol

4-Chlorophenylhydrazine hydrochloride is used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorophenylhydrazine hydrochloride is used as a pharmaceutical intermediates.

Solubility
Soluble in hot water, methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 1073-70-7
Molecular Formula C6H8Cl2N2
Molecular Weight (g/mol) 179.044
MDL Number MFCD00012943
InChI Key YQVZREHUWCCHHX-UHFFFAOYSA-N
Synonym 4-chlorophenylhydrazine hydrochloride, 4-chlorophenyl hydrazine hydrochloride, p-chlorophenylhydrazine hydrochloride, p-chlorophenylhydrazine hcl, 4-chlorophenylhydrazine hcl, 1-4-chlorophenyl hydrazine hydrochloride, 4-chloro-phenyl-hydrazine hydrochloride, chlorophenylhydrazine 4-hcl, hydrazine, 4-chlorophenyl-, monohydrochloride, 4-chlorophenylhydrazine, chloride
PubChem CID 71600
IUPAC Name (4-chlorophenyl)hydrazine;hydrochloride
SMILES C1=CC(=CC=C1NN)Cl.Cl

Specifications

Melting Point ∼216°C (decomposition)
Odor Odorless
Quantity 100 g
Beilstein 3596254
Solubility Information Soluble in hot water,methanol.
Formula Weight 179.05
Percent Purity 97%
Chemical Name or Material 4-Chlorophenylhydrazine hydrochloride
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H350-H315-H319-H335
Toxic if swallowed.
May cause cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 214-030-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only