accessibility menu, dialog, popup

UserName

CdOHNOOOHNOO

CAS RN 10325-94-7

IUPAC Name: cadmium(2+) bis(nitric acid)
Synonyms: cadmium nitrate
Molecular Weight (g/mol): 238.44
Molecular Formula: CdH2N2O6
InChi Key: IZIPRBSCGMISKX-UHFFFAOYSA-N
SMILES: [Cd++].O[N+]([O-])=O.O[N+]([O-])=O
Grade 
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results