missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Cadmium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical

Catalog No. RV010270500
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010270500 500 mL
RV010270100 100 mL
RV010270250 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RV010270500 Supplier Ricca Chemical Company Supplier No. RV010270500N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 10325-94-7 , 7697-37-2 , 7732-18-5
Molecular Formula CdN2O6
Molecular Weight (g/mol) 236.422
InChI Key XIEPJMXMMWZAAV-UHFFFAOYSA-N
PubChem CID 25154
ChEBI CHEBI:77732
IUPAC Name cadmium(2+);dinitrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Cd+2]

Specifications

Packaging Natural Poly Bottle
CAS Min % 95.83%, 1.96%, 0.21%
CAS Max % 99.75%, 2.04%, 0.21%
Specific Gravity 1
Physical Form Liquid
Melting Point Approximately 0°C
Type Standard
Quantity 500 mL
Grade High Purity