missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium Standard for AAS, 1,000 mg/L Cd in Nitric Acid, MilliporeSigma™ Supelco™
Specifications
Specifications
| Product Line | TraceCERT |
| Concentration | 1000 mg/L Cd in nitric acid |
| Quantity | 250 mL |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC