CAS RN 10294-41-4
Cerium(III) nitrate hexahydrate, REacton™, 99.5% (REO)
CAS: 10294-41-4 Molecular Formula: CeH12N3O15 Molecular Weight (g/mol): 434.22 MDL Number: MFCD00149631 InChI Key: QQZMWMKOWKGPQY-UHFFFAOYSA-N Synonym: cerium iii nitrate hexahydrate,cerium nitrate hexahydrate,cerous nitrate hexahydrate,unii-fl1r38jebb,fl1r38jebb,nitric acid cerium salt,cerium iii nitratehexahydrate,ce.3no3.6h2o,ksc494m5f PubChem CID: 16211466 IUPAC Name: cerium(3+) hexahydrate trinitrate SMILES: O.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Cerium(III) nitrate hexahydrate, 99.5%
CAS: 10294-41-4 Molecular Formula: CeH12N3O15 Molecular Weight (g/mol): 434.22 MDL Number: MFCD00149631 InChI Key: QQZMWMKOWKGPQY-UHFFFAOYSA-N Synonym: cerium iii nitrate hexahydrate,cerium nitrate hexahydrate,cerous nitrate hexahydrate,unii-fl1r38jebb,fl1r38jebb,nitric acid cerium salt,cerium iii nitratehexahydrate,ce.3no3.6h2o,ksc494m5f PubChem CID: 16211466 SMILES: O.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Cerium ICP Standard, 1000 ppm Ce in 3% HNO3, Ricca Chemical
CAS: 10294-41-4 Molecular Formula: CeH12N3O15 Molecular Weight (g/mol): 434.22 MDL Number: MFCD00149631 InChI Key: QQZMWMKOWKGPQY-UHFFFAOYSA-N PubChem CID: 16211466 IUPAC Name: cerium(3+) hexahydrate trinitrate SMILES: O.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
VeriSpec™ Cerium Standard for ICP/MS 10 ppm in 2% HNO3, Ricca Chemical
CAS: 10294-41-4 Molecular Formula: CeH12N3O15 Molecular Weight (g/mol): 434.22 MDL Number: MFCD00149631 InChI Key: QQZMWMKOWKGPQY-UHFFFAOYSA-N PubChem CID: 16211466 IUPAC Name: cerium(3+) hexahydrate trinitrate SMILES: O.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Cerium AA Standard, 1000 ppm Ce in 3% HNO3, Ricca Chemical
CAS: 10294-41-4 Molecular Formula: CeH12N3O15 Molecular Weight (g/mol): 434.218 InChI Key: QQZMWMKOWKGPQY-UHFFFAOYSA-N PubChem CID: 16211466 IUPAC Name: cerium(3+);trinitrate;hexahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Ce+3]
VeriSpec™ Cerium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical
CAS: 10294-41-4 Molecular Formula: CeH12N3O15 Molecular Weight (g/mol): 434.22 MDL Number: MFCD00149631 InChI Key: QQZMWMKOWKGPQY-UHFFFAOYSA-N PubChem CID: 16211466 IUPAC Name: cerium(3+) hexahydrate trinitrate SMILES: O.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Cerium ICP Standard, 10,000 ppm Ce in 10% HNO3, Ricca Chemical
CAS: 10294-41-4 Molecular Formula: CeH12N3O15 Molecular Weight (g/mol): 434.218 InChI Key: QQZMWMKOWKGPQY-UHFFFAOYSA-N PubChem CID: 16211466 IUPAC Name: cerium(3+);trinitrate;hexahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Ce+3]
VeriSpec™ Cerium (Ce) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical
CAS: 10294-41-4 Molecular Formula: CeH12N3O15 Molecular Weight (g/mol): 434.22 MDL Number: MFCD00149631 InChI Key: QQZMWMKOWKGPQY-UHFFFAOYSA-N PubChem CID: 16211466 IUPAC Name: cerium(3+) hexahydrate trinitrate SMILES: O.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
VeriSpec™ Cerium Standard for ICP/MS 100 ppm in 2% HNO3, Ricca Chemical
CAS: 10294-41-4 Molecular Formula: CeH12N3O15 Molecular Weight (g/mol): 434.22 MDL Number: MFCD00149631 InChI Key: QQZMWMKOWKGPQY-UHFFFAOYSA-N PubChem CID: 16211466 IUPAC Name: cerium(3+) hexahydrate trinitrate SMILES: O.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O