missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerium(III) nitrate hexahydrate, 99.5%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless to White |
| Physical Form | Crystalline Powder or Agglomerating Crystals |
| Quantity | 500 g |
| Packaging | Plastic bottle |
| Assay Percent Range | 99.4% min. (CeO2/TREO basis) |
| Linear Formula | Ce(NO3)3·6H2O |
| Fieser | 16,67 |
| Merck Index | 10,1959 |
| Solubility Information | Solubility in water: 1754g/L (25°C). Other solubilities: soluble in alcohol and acetone |
| IUPAC Name | cerium(3+);trinitrate;hexahydrate |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May intensify fire; oxidizer. Causes serious eye damage. Very toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Take any precaution to avoid mixing with combustibles. In case of major |
| RTECSNumber | FK6300000 |
RUO – Research Use Only