missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Cerium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical

Catalog No. RV010271500
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010271500 500 mL
RV010271100 100 mL
RV010271250 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RV010271500 Supplier Ricca Chemical Company Supplier No. RV010271500N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 10294-41-4 , 7697-37-2 , 7732-18-5
Molecular Formula CeH12N3O15
Molecular Weight (g/mol) 434.22
MDL Number MFCD00149631
InChI Key QQZMWMKOWKGPQY-UHFFFAOYSA-N
PubChem CID 16211466
IUPAC Name cerium(3+) hexahydrate trinitrate
SMILES O.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Natural Poly Bottle
CAS Min % 95.74%, 1.96%, 0.30%
CAS Max % 99.64%, 2.04%, 0.32%
Specific Gravity 1
Physical Form Liquid
Melting Point Approximately 0°C
Type Standard
Quantity 500 mL
Grade High Purity