missing translation for 'onlineSavingsMsg'
Learn More

Cerium AA Standard, 1000 ppm Ce in 3% HNO3, Ricca Chemical

Catalog No. p-3251476
Change view
Click to view available options
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
ACE1KN100 Natural LDPE Bottle
ACE1KN500 Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ACE1KN100 Supplier Ricca Chemical Company Supplier No. ACE1KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10294-41-4 , 7697-37-2 , 7732-18-5
Molecular Formula CeH12N3O15
Molecular Weight (g/mol) 434.218
InChI Key QQZMWMKOWKGPQY-UHFFFAOYSA-N
PubChem CID 16211466
IUPAC Name cerium(3+);trinitrate;hexahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Ce+3]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Ce
CAS Min % 93.0, 4.21, 0.29
CAS Max % 97.0, 4.39, 0.3
Equivalence 1 mL = 1 mg Ce
Chemical Name or Material Cerium AA Standard