missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) nitrate hexahydrate, REacton™, 99.5% (REO)

Catalog No. AA11329A1
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11329A1 1 kg
AA1132909 10 g
AA1132922 100 g
AA1132936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA11329A1 Supplier Thermo Scientific Chemicals Supplier No. 011329A1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Separation of Cerium From Other Rare Earths, Catalyst For Hydrolysis of Phosporic Acid Esters | CAS: 10294-41-4 | CeH12N3O15 | 434.22 g/mol

Cerium(III) nitrate hexahydrate is used in the separation of cerium from other rare earths. It plays an important role as a catalyst for hydrolysis of phosphoric acid esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10294-41-4
Molecular Formula CeH12N3O15
Molecular Weight (g/mol) 434.22
MDL Number MFCD00149631
InChI Key QQZMWMKOWKGPQY-UHFFFAOYSA-N
Synonym cerium iii nitrate hexahydrate, cerium nitrate hexahydrate, cerous nitrate hexahydrate, unii-fl1r38jebb, fl1r38jebb, nitric acid cerium salt, cerium iii nitratehexahydrate, ce.3no3.6h2o, ksc494m5f
PubChem CID 16211466
IUPAC Name cerium(3+) hexahydrate trinitrate
SMILES O.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Physical Form Crystalline Aggregates
Quantity 1 kg
Sensitivity Hygroscopic
Assay Percent Range 99.5%
Solubility Information Soluble in water,alcohol and acetone.
Formula Weight 434.23 (326.13 Anhydrous)
Chemical Name or Material Cerium(III) nitrate hexahydrate, hexahydrate
Hazard Category H272-H315-H318-H350
Precautionary Statement P201-P202-P220-P221-P264b-P271-P280-P302+P352-P305+P351+P338-P308+P313-P310-P332+P313-P362-P370+P380+P375-P372-P501c
DOTInformation DOT Class: 5.1, Packaging Group: III
EINECSNumber 233-297-2
RTECSNumber FK6300000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only