missing translation for 'onlineSavingsMsg'
Learn More

Cerium ICP Standard, 1000 ppm Ce in 3% HNO3, Ricca Chemical

Catalog No. PCE1KN500
Change view
Click to view available options
Package: Natural Poly; Size: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PCE1KN500 Package: Natural Poly; Size: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PCE1KN500 Supplier Ricca Chemical Company Supplier No. PCE1KN500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Natural Poly Bottle
CAS 10294-41-4, 7697-37-2, 7732-18-5
Concentration or Composition Notes 1,000ppm Ce
CAS Min % 93.0, 4.21, 0.29
CAS Max % 97.0, 4.39, 0.3
Equivalence 1 mL = 1 mg Ce
Molecular Formula CeH12N3O15
InChI Key QQZMWMKOWKGPQY-UHFFFAOYSA-N
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Ce+3]
IUPAC Name cerium(3+);trinitrate;hexahydrate
Molecular Weight (g/mol) 434.218
PubChem CID 16211466
Chemical Name or Material Cerium ICP Standard
Show More Show Less