accessibility menu, dialog, popup

UserName

SrONOOONOO

CAS RN 10042-76-9

IUPAC Name: strontium(2+) dinitrate
Synonyms: strontium nitrate
Molecular Weight (g/mol): 211.63
Molecular Formula: N2O6Sr
InChi Key: DHEQXMRUPNDRPG-UHFFFAOYSA-N
SMILES: [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Grade 
  • (6)
  • (6)
  • (2)
  • (2)
  • (2)
  • (2)
  • (10)
  • (1)
Physical Form 
  • (5)
  • (4)
  • (3)
  • (6)
  • (3)
  • (2)
  • (2)
Quantity 
  • (4)
  • (4)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (7)
  • (2)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (6)
  • (2)
  • (2)
  • (18)
  • (6)
  • (17)
  • (6)
  • (6)
  • (2)
  • (2)
  • (2)
  • (2)
  • (10)
  • (1)
  • (5)
  • (4)
  • (3)
  • (6)
  • (3)
  • (2)
  • (2)
  • (4)
  • (4)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (7)
  • (2)
  • (6)
  • (2)