missing translation for 'onlineSavingsMsg'
Learn More

Strontium AA Standard, 1000 ppm Sr in 3% HNO3, Ricca Chemical

Catalog No. p-3255655
Change view
Click to view available options
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
ASR1KN100 Natural LDPE Bottle
ASR1KN500 Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ASR1KN100 Supplier Ricca Chemical Company Supplier No. ASR1KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10042-76-9 , 7697-37-2 , 7732-18-5
Molecular Formula N2O6Sr
Molecular Weight (g/mol) 211.63
MDL Number MFCD00011248
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
PubChem CID 24848
IUPAC Name strontium(2+) dinitrate
SMILES [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Sr
CAS Min % 94.0, 4.22, 0.23
CAS Max % 97.0, 4.39, 0.24
Equivalence 1 mL = 1 mg Sr
Chemical Name or Material Strontium AA Standard