missing translation for 'onlineSavingsMsg'
Learn More

Strontium ICP Standard, 10,000 ppm Sr in 5% HNO3, Ricca Chemical

Catalog No. PSR10KN100
Change view
Click to view available options
Package: Natural LDPE; Size: 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PSR10KN100 Package: Natural LDPE; Size: 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PSR10KN100 Supplier Ricca Chemical Company Supplier No. PSR10KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Natural LDPE Bottle
CAS 10042-76-9, 7697-37-2, 7732-18-5
CAS Min % 89.0, 6.7, 2.15
Concentration or Composition Notes 10,000ppm Sr
CAS Max % 93.0, 6.97, 2.24
Equivalence 1 mL = 10 mg Sr
Molecular Formula N2O6Sr
MDL Number MFCD00011248
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
SMILES [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
IUPAC Name strontium(2+) dinitrate
Molecular Weight (g/mol) 211.63
PubChem CID 24848
Chemical Name or Material Strontium ICP Standard
Show More Show Less