missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium nitrate, 99+%, ACS reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 570.0°C |
| Color | White |
| pH | 5.0 to 7.0 (5% soln. at 25°C) |
| Physical Form | Crystals |
| Quantity | 500 g |
| Loss on Drying | 0.1% max. (105°C) |
| Packaging | Plastic bottle |
| Assay Percent Range | 99% min. (Complexometry) |
| Density | 2.99 |
| Linear Formula | Sr(NO3)2 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May intensify fire; oxidizer. Causes serious eye damage. Harmful if swallowed. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Take any precaution to avoid mixing with combustibles. In case of major |
| EINECSNumber | 233-131-9 |
| RTECSNumber | WK9800000 |
| TSCA | TSCA |
RUO – Research Use Only