missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Standard, 1000mg/L in 2% HNO3 (Assurance Grade), SPEX CertiPrep™
$143.43 - $376.50
Chemical Identifiers
| CAS | 7440-66-6 |
|---|---|
| Molecular Formula | Zn |
| Molecular Weight (g/mol) | 65.38 |
| MDL Number | MFCD00011291 |
| InChI Key | HCHKCACWOHOZIP-UHFFFAOYSA-N |
| PubChem CID | 23994 |
| ChEBI | CHEBI:30185 |
| IUPAC Name | zinc |
| SMILES | [Zn] |
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Zn | 1000μg/mL |
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Chemical Identifiers
| 7440-66-6 | |
| 65.38 | |
| HCHKCACWOHOZIP-UHFFFAOYSA-N | |
| CHEBI:30185 | |
| [Zn] |
| Zn | |
| MFCD00011291 | |
| 23994 | |
| zinc |
Specifications
| Triple-leached LDPE bottle | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 1.0,5.0,94.0 | |
| MFCD00011291 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| HCHKCACWOHOZIP-UHFFFAOYSA-N | |
| 2% HNO3 | |
| 65.38 | |
| 23994 | |
| Plasma | |
| Zinc Standard |
Safety and Handling
May cause cancer.
DOTInformation : Nonhazardous
Recommended Storage : Ambient