missing translation for 'onlineSavingsMsg'
Learn More

Zinc Standard, 1000mg/L in 2% HNO3 (Assurance Grade), SPEX CertiPrep™

Supplier:  SPEX CertiPrep Group LLC PLZN22T

 View more versions of this product

Catalog No. PLZN22T


Only null left
Explore available promotions

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Zinc Standard
  • Made with ASTM Type I Water
  • 1,000 μg/mL concentration
  • 250 mL volume

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Specifications

Specifications

Zinc Standard
7440-66-6 , 7697-37-2 , 7732-18-5
1.0,5.0,94.0
Zn
AA, ICP
HCHKCACWOHOZIP-UHFFFAOYSA-N
zinc
250 mL
CHEBI:30185
Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLZN2-2Y, -2T, -2X    10,000    PLZN2-3Y, -3X
2-5% HCl    1000μg/mL    PLZN1-2X    10,000    PLZN1-3X
Poly Bottle
Assurance Certificate of Analysis reports up 30 elements at ppb levels.
1.0,5.0,94.0
MFCD00011291
Certified Reference Material
[Zn]
65.38
23994
Plasma

Chemical Identifiers

7440-66-6
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
CHEBI:30185
[Zn]
Zn
MFCD00011291
23994
zinc
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

May cause cancer.

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions