missing translation for 'onlineSavingsMsg'
Learn More

Zinc Standard, 1000mg/L in 2% HNO3 (Assurance Grade), SPEX CertiPrep™

Supplier:  SPEX CertiPrep Group LLC PLZN22M

 View more versions of this product

Catalog No. PLZN22M


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Zn1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Zinc Standard
7440-66-6 , 7697-37-2 , 7732-18-5
1.0,5.0,94.0
Zn
AA, ICP
Certified Reference Material
[Zn]
zinc
30 mL
CHEBI:30185
Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLZN2-2Y, -2T, -2X    10,000    PLZN2-3Y, -3X
2-5%
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
1.0,5.0,94.0
MFCD00011291
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
HCHKCACWOHOZIP-UHFFFAOYSA-N
2% HNO3
65.38
23994
Plasma

Chemical Identifiers

7440-66-6
65.38
HCHKCACWOHOZIP-UHFFFAOYSA-N
CHEBI:30185
[Zn]
Zn
MFCD00011291
23994
zinc
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

May cause cancer.

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions