missing translation for 'onlineSavingsMsg'
Learn More

Zinc chloride, 2M solution in 2-MeTHF, AcroSeal™

CAS: 7646-85-7 | Cl2Zn | 136.29 g/mol

284,04 $ - 284,04 $

Spécifications

Chemical Name or Material Zinc chloride
Name Note 2M solution in 2-Methyltetrahydrofuran
CAS 7646-85-7,96-47-9
Color Colorless
Physical Form Liquid
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
AC428561000
Afficher les documents
Thermo Scientific Chemicals
428561000
100 mL AcroSeal™ Glass Bottle N/A
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

Zinc chloride
7646-85-7,96-47-9
Liquid
ZnCl2
−11°C
zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
Cl[Zn]Cl
136.29
CHEBI:49976
23 to 27%
1.0700g/mL
2M solution in 2-Methyltetrahydrofuran
Colorless
Cl2Zn
MFCD00011295
15, 10331
JIAARYAFYJHUJI-UHFFFAOYSA-L
dichlorozinc
5727
136.29
1.07
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only