missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zero Oxygen Standard, for Calibration of Dissolved Oxygen Meters, Ricca Chemical
$145.19 - $251.78
Chemical Identifiers
| CAS | 7791-13-1 |
|---|---|
| Molecular Formula | Cl2CoH12O6 |
| Molecular Weight (g/mol) | 237.92 |
| MDL Number | MFCD00149652 |
| InChI Key | GFHNAMRJFCEERV-UHFFFAOYSA-L |
| IUPAC Name | λ2-cobalt(2+) hexahydrate dichloride |
| SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Co++] |
Chemical Identifiers
| 7791-13-1 | |
| 237.92 | |
| GFHNAMRJFCEERV-UHFFFAOYSA-L | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Co++] |
| Cl2CoH12O6 | |
| MFCD00149652 | |
| λ2-cobalt(2+) hexahydrate dichloride |
Specifications
| Natural Poly Bottle | |
| 93.0,4.67,0.0023 | |
| Liquid | |
| MFCD00149652 | |
| Approximately 0°C | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Co++] | |
| 237.92 | |
| Zero Oxygen Standard, For Calibration of Dissolved Oxygen Meters |