Learn More
DWK Life Sciences Wheaton™ Peroxide Cured Silicone Tubing For General Lab Use
Assembly consists of 1.5m (5 ft.) section of silicone tubing with a wall thickness of 2.3mm, a borosilicate glass sinker, and a polypropylene delivery nozzle.
$147.71 - $541.36
Specifications
| Material | Silicone |
|---|---|
| Thickness (English) | 0.09 in. |
| Thickness (Metric) | 2.3 mm |
| Catalog Number | Mfr. No. | Diameter (Metric) Inner | Overall Length (Metric) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (Metric) Inner | Overall Length (Metric) | Price | Quantity & Availability | ||||
|
02913310
|
DWK Life Sciences
374308 |
8 mm | 760 cm |
Case for $541.36
|
|
||||
|
1368310
|
DWK Life Sciences
374314 |
8 mm | 150 cm |
Each for $215.40
|
|
||||
|
136839
|
DWK Life Sciences
374313 |
6 mm | 150 cm |
Each for $201.57
|
|
||||
|
136838
|
DWK Life Sciences
374311 |
3 mm | 150 cm |
Each for $147.71
|
|
||||
|
02928101
|
DWK Life Sciences
374304 |
2 mm | 760 cm |
Case for $237.52
|
|
||||
|
22246640
|
DWK Life Sciences
374310 |
2 mm | 150 cm |
Each for $223.57
|
|
||||
|
034108D
|
DWK Life Sciences
374305 |
3 mm | 760 cm |
Pack for $407.68
|
|
||||
|
034108E
|
DWK Life Sciences
374306 |
6 mm | 760 cm |
Pack for $541.36
|
|
||||
Description
Assembly consists of 1.5m (5 ft.) section of silicone tubing with a wall thickness of 2.3mm, a borosilicate glass sinker, and a polypropylene delivery nozzle
Specifications
| Silicone | |
| 0.09 in. | |
| 2.3 mm |