Learn More
DWK Life Sciences Wheaton™ Peroxide Cured Silicone Tubing For General Lab Use
Assembly consists of 1.5m (5 ft.) section of silicone tubing with a wall thickness of 2.3mm, a borosilicate glass sinker, and a polypropylene delivery nozzle.
Supplier:  DWK Life Sciences 374306
 View more versions of this product
Catalog No. 034108E
Only null left
Description
Description
  • Autoclavable
Specifications
Chemical Identifiers
Silicone
0.09 in.
2.3 mm
Specifications
23/100 in.
Silicone
760 cm
2.3 mm
Tubing
6 mm
25 ft.
0.09 in.
Peristaltic pump
Product Suggestions
Product Suggestions