Learn More
DWK Life Sciences Wheaton™ Peroxide Cured Silicone Tubing For General Lab Use
Assembly consists of 1.5m (5 ft.) section of silicone tubing with a wall thickness of 2.3mm, a borosilicate glass sinker, and a polypropylene delivery nozzle.
Supplier:  DWK Life Sciences 374314
 View more versions of this product
Catalog No. 1368310
Only null left
Description
Description
  • Autoclavable
Specifications
Chemical Identifiers
Silicone
0.09 in.
2.3 mm
Specifications
31/100 in.
Silicone
150 cm
2.3 mm
Sterile
Tubing Assembly
8 mm
5 ft.
0.09 in.
Peristaltic pump
Borosilicate Glass sinker on one end and Polypropylene delivery nozzle on the other end
Product Suggestions
Product Suggestions