Learn More
DWK Life Sciences Wheaton™ Impellers for Celstir™ Spinner Flasks
Replacement for Double Sidearm Celstir and Jacketed Double Sidearm Celstir.
Supplier:  DWK Life Sciences 356904
 View more versions of this product
Catalog No. 06-416-5F
Only null left
Explore available promotions
Description
Description
  • This non-breakable stainless steel shaft, for replacing the glass shaft included with Celstir™ flasks, may be retrofitted to the existing Celstir flasks.
Specifications
Specifications
1000mL Celstir Spinner Flask
Top cap with liners, glass rod, magnet holder, magnet and stirring paddle
Impeller
Product Suggestions
Product Suggestions