Learn More
DWK Life Sciences Wheaton™ Impellers for Celstir™ Spinner Flasks
Replacement for Double Sidearm Celstir and Jacketed Double Sidearm Celstir.
Supplier: DWK Life Sciences 356902
Description
- This non-breakable stainless steel shaft, for replacing the glass shaft included with Celstir™ flasks, may be retrofitted to the existing Celstir flasks.
Spécifications
| 500mL Celstir Spinner Flask | |
| Top cap with liners, glass rod, magnet holder, magnet and stirring paddle | |
| Impeller |