Learn More
DWK Life Sciences Wheaton™ HDPE Serum Bottles
Wheaton Serum bottle is manufactured from HDPE and widely used for animal health products and storage of non-medical products.
Supplier:  DWK Life Sciences 224036
 Afficher plus de versions de ce produit
Numéro de catalogue. 06447X
Il en reste null
Description
Description
  • Manufactured from high density polyethylene
  • Widely used for animal health products and storage of non-medical products
  • Accepts standard rubber stoppers and aluminum seals
Spécifications
Identifiants chimiques
Natural
20 mm
HDPE
Rubber
0.78 in.
13 mm
Serum Bottle
Spécifications
2.02 oz., 60 mL
2.02 oz.
1.49 in.
20 mm
HDPE
Serum Bottle
Rubber
60 mL
Natural
0.78 in.
13 mm
84 mm
3.3 in.
38 mm
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS