missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ HDPE Serum Bottles
Wheaton Serum bottle is manufactured from HDPE and widely used for animal health products and storage of non-medical products.
$499.30 - $664.68
Specifications
| Color | Natural |
|---|---|
| Diameter (English) Inner Neck | 0.78 in. |
| Diameter (Metric) Outer Neck | 20 mm |
| Diameter (Metric) Inner Neck | 13 mm |
| Material | HDPE |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
06446W
|
DWK Life Sciences
224031 |
15 mL |
Case of 300 Each for $630.76
|
|
|||||
|
06446V
|
DWK Life Sciences
224033 |
30 mL |
Case of 200 Each for $664.68
|
|
|||||
|
06447X
|
DWK Life Sciences
224036 |
60 mL |
Case of 100 Each for $499.30
|
|
|||||
|
06448T
|
DWK Life Sciences
224037 |
120 mL |
Case of 100 Each for $536.27
|
|
|||||
Description
- Manufactured from high density polyethylene
- Widely used for animal health products and storage of non-medical products
- Accepts standard rubber stoppers and aluminum seals
Specifications
| Natural | |
| 20 mm | |
| HDPE | |
| Rubber |
| 0.78 in. | |
| 13 mm | |
| Serum Bottle |