Learn More
DWK Life Sciences Wheaton™ Connection With Black Phenolic Caps
The Wheaton Connection™ Screw Thread Connector Safely joins two exterior threaded glass components without hooks, springs or clamps. Screw cap configuration creates a grease-free, vacuum-tight seal.
Supplier:  DWK Life Sciences 125496
 Afficher plus de versions de ce produit
Numéro de catalogue. 02-912-543
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Inert contact surface
  • Easy to assemble
  • Connection can be used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
  • Phenolic connectors have a working temperature of up to 180°C
  • Autoclavable
Spécifications
Spécifications
Used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
Connector
13-425, 24-400
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS