Learn More
Thermo Scientific Chemicals Vinblastine Sulfate, 98%
Anticancer agent; microtubule disrupter; Induces apoptosis | CAS: 143-67-9 | C46H60N4O13S, C46H60N4O13S | 909.06 g/mol
Supplier: Thermo Scientific Chemicals J63598MA
| Quantity | 10 mg |
|---|
Description
Anticancer agent; microtubule disrupter; Induces apoptosis.Vinblastine sulfate is therapeutically used as an antineoplastic. It is also used in the treatment of Hodgkin disease, choriocarcinoma, acute and chronic leukemias.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 143-67-9 | |
| C46H60N4O13S | |
| Vincaleukblastine sulfate |
Spécifications
| 10 mg | |
| 2°C to 8°C | |
| 98% | |
| C46H60N4O13S | |
| Vincaleukblastine sulfate | |
| Soluble in water or DMSO | |
| Powder |
| White | |
| Anticancer agent; microtubule disrupter; Induces apoptosis | |
| 143-67-9 | |
| 909.06g/mol | |
| 267°C | |
| Signal Transduction Reagent | |
| 3659812 |
Sécurité et manipulation
EINECSNumber : 205-606-0
Recommended Storage : Keep cold