missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ ViiA™ 7 Dx Block Upgrades

Front access makes it easy to change thermal cycling block formats without having to move attached peripherals such as robots or computers.

$20,760.00 - $34,380.00

Spécifications

For Use With (Equipment) ViiA™ 7 Dx System
Product Type Block Upgrade
For Use With (Application) Real time PCR (qPCR)
voir les spécifications

Comprend: Includes 384-well Dx Block, heated cover, plate adaptor and calibration kit.

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Description Content And Storage Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Quantity Description Content And Storage Prix Quantity & Availability  
4453549
Encompass
Afficher les documents Détails de la promotion
Applied Biosystems™
4453549
1 Upgrade, 1 x 384-well Plate 384-well Plate 384-well Dx Block, heated cover, plate adaptor and calibration kit. N/A
N/A
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
4453550
Encompass
Afficher les documents Détails de la promotion
Applied Biosystems™
4453550
1 Upgrade, 1 Array Card Array Card Array Card Dx Block, heated cover, plate adaptor and calibration kit. N/A
N/A
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Description

Description

The ViiA™ 7 Dx Instrument puts high-productivity qPCR at your fingertips. With the new OptiFlex™ optical system and 4 interchangeable block formats from 96-well to 384-well to Micro Fluidic Array Card, the ViiA™ 7 Dx Instrument is available for in-vitro diagnostics use.

Reaction Speed: Standard
Spécifications

Spécifications

ViiA™ 7 Dx System
Block Upgrade
Real time PCR (qPCR)
Videos
FDS
Documents

Documents