missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ ViiA™ 7 Dx Block Upgrades
Description
Includes
Includes 384-well Dx Block, heated cover, plate adaptor and calibration kit.
The ViiA™ 7 Dx Instrument puts high-productivity qPCR at your fingertips. With the new OptiFlex™ optical system and 4 interchangeable block formats from 96-well to 384-well to Micro Fluidic Array Card, the ViiA™ 7 Dx Instrument is available for in-vitro diagnostics use.
Reaction Speed: Standard
Specifications
Specifications
| Content And Storage | Array Card Dx Block, heated cover, plate adaptor and calibration kit. |
| For Use With (Equipment) | ViiA™ 7 Dx System |
| Product Type | Block Upgrade |
| For Use With (Application) | Real time PCR (qPCR) |
| Description | Array Card |
| Quantity | 1 Upgrade, 1 Array Card |